Phthalates Polyethylene Standard in Medium Density Polyethylene, SPEX CertiPrep
Catalog # SXCRM-PE001
Supplier: SPEX CERTIPREP LLC
CAS Number:
Specifications
- Concentration:5 g
- Matrix:Polyethylene (medium density)
- Pack type:Glass bottle, amber
- Description:Phthalates Polyethylene Standard in Medium Density Polyethylene
- Size:5 g
- Cat. No.:SXCRM-PE001
Specifications
About this item
Consumer safety standards, phthalates in polyethylene QC standards.
Polyethylene (PE) is one of the world’s most common plastics. PE is used in a variety of common consumer products including children’s toys and care items. Current US regulations limit the concentrations of certain phthalates in childcare articles and children’s toys. Laboratories are tasked with the analysis of children’s toys for these potentially hazardous phthalates.