Order Entry
United States
ContactUsLinkComponent
Sodium hydroxide concentrate 1.25 N, SimpleDist Snip & Pour™ Reagent
Sodium hydroxide concentrate 1.25 N, SimpleDist Snip & Pour™ Reagent
Catalog # 76436-850
CAS Number:  
undefined
Sodium hydroxide concentrate 1.25 N, SimpleDist Snip & Pour™ Reagent
Catalog # 76436-850
Supplier Number:  C7115
CAS Number:  

Specifications

  • Pack type:
    PTFE tube
  • Size:
    50×5 ml
  • Cat. No.:
    76436-850

Specifications

About this item

 :
 :40 g/mol
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548