
Sulfuric acid concentrate 0.2 N, Snip & Pour™
Catalog # 76436-846
Supplier: Environmental Express
CAS Number:
Specifications
- Pack type:PFA tube
- Size:50×10 ml
- Cat. No.:76436-846
Specifications
About this item
:
:98.08
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589