
Lithium carbonate, flux additive, ultrapure grade for XRF (X-ray fluorescence), for ICP and AAS
Catalog # SXFFB-401-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:LDPE bottle
- Size:1 kg
- Cat. No.:SXFFB-401-03
Specifications
About this item
:
:73.89 g/mol
:GHS07
CAS Number:554-13-2
Formula:Li₂CO₃
MDL Number:MFCD00011084