Lithium metaborate : lithium bromide (99.5% : 0.5%), fusion flux, ultrapure grade for XRF (X-ray fluorescence), for ICP and AAS
Catalog # SXFFB-0005-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:LDPE bottle
- Size:1 kg
- Cat. No.:SXFFB-0005-03
Specifications
About this item
:
:GHS07
:13453-69-5