Fast Track Diagnostics Real-Time PCR Multiplexing Kits, Siemens Healthineers
Supplier: Siemens Healthineers
Certificates
About this item
Kits identify a wide range of viruses, bacteria, fungi and parasites. By detecting up to four pathogens in one tube, and up to thirty-three pathogens in a single sample, each assay provides labs quick access to the detailed data you need for your research studies.
- One-step real-time PCR multiplexing kits for the simultaneous detection of viruses, bacteria, fungi or parasites
- Compatible for use on many commercially available Real Time-PCR platforms. Refer to the Fast Track Diagnostic website for a detailed list
- Prior to shipping, a signed letter is required acknowledging that these products will be used for research use only
The components of the FTD product should be stored in the original packaging between –30 and –10 °C and are stable until the expiration date stated on the label. Freeze the product immediately after use. Note: Repeated freeze/thaw of reagents may reduce sensitivity.
Caution: Products are for laboratory research only and are not intended for human clinical diagnostic, prognostic, or monitoring use.
Frequently Bought Together