To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Concentration:1000 µg/ml
- Matrix:Base Oil 20
- Pack type:LDPE bottle
- Size:50 g
- Environmentally Preferable:
- Cat. No.:SXORG-SB8-2Z
- Element:Antimony
Organometallic single-element oil standards are guaranteed stable and accurate, together with a wide range of applications and features:
- Wear metals
- Crude oils
- Additive metals
- Environmental monitoring Petrochemical testing
- Pharmaceuticals
- Food processing
- Sulfur in diesel fuel
The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. The examples are endless. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.