ICP-MS Spike Sample 3, SPEX CertiPrep
Product Details & Documents
CLP spike standard for ICP & ICP-MS.
Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.
The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by SPEX CertiPrep for convenience of use and stability.
US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentrationlevels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:
- Exhibit C, Inorganic Target Analyte List (TAL)
- Exhibit D, Analytical Methods
- Exhibit E, QA/QC Requirements
CLP ILM 05.2 Standards for ICP-MS
Spike standard, CL-SPIKE-3, provides all of the analytes required for the ICP and AA spike.
Aluminum200 µg/mL
Barium200 µg/mL
Cobalt50 µg/mL
Manganese50 µg/mL
Nickel50 µg/mL
Vanadium50 µg/mL
Zinc50 µg/mL
Copper25 µg/mL
Chromium20 µg/mL
Antimony10 µg/mL
Beryllium5 µg/mL
Cadmium5 µg/mL
Silver5 µg/mL
Thallium5 µg/mL
Arsenic4 µg/mL
Lead2 µg/mL
Selenium1 µg/mL
Specifications
Specifications
- Matrix:5% HNO₃/tr. Tartaric acid/tr. HF
- Pack type:LDPE bottle
- Description:ICP-MS spike sample 3
- Size:125 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



