Avantor® Hichrom HI-SOLVE 1, GC Columns
Supplier: VWR International
Certificates
About this item
Application specifc columns developed for complex solvents mixtures analysis. HI-SOLVE 1 is ideal for aromatics and oxygenates in gasoline.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76429-096
- Phase name:
- HI-SOLVE 1
- Phase composition:
- Proprietary Phase developed for complex solvent mix