Order Entry
United States
ContactUsLinkComponent
 
Claritas PPT® Grade ICP-MS Multi-Element Solution 5, SPEX CertiPrep
Catalog # SXCLMS-5
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Claritas PPT® Grade ICP-MS Multi-Element Solution 5, SPEX CertiPrep
Catalog # SXCLMS-5
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CLMS-5
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    LDPE bottle
  • Description:
    ICP-MS Multi-Element Solution 5
  • Size:
    125 ml
  • Cat. No.:
    SXCLMS-5

Specifications

About this item

Claritas PPT® multi-element standards for ICP-MS.

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.

The Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.

Multi-Solution Solution Standards: Designed to contain virtually every element in the mass spectrum for concentration verification checks.

Certifications: SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.