Specifications
- Concentration:10 µg/ml
- Matrix:5% HNO₃
- Pack type:LDPE bottle
- Description:ICP-MS Multi-Element Solution 2A without Mercury
- Size:125 ml
- Cat. No.:SXCLMS-2AN
Specifications
About this item
Claritas PPT® multi-element standards for ICP-MS.
SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.
The Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.
Multi-Solution Solution Standards: Designed to contain virtually every element in the mass spectrum for concentration verification checks.