Specifications
- Concentration:10 µg/ml
- Matrix:2% HNO₃
- Pack type:LDPE bottle
- Description:ICP-MS Internal Standard 1
- Size:125 ml
- Cat. No.:SXCLISS-1
Specifications
About this item
Claritas PPT® Multi-Element Standards for ICP-MS.
SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.
Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.
May be used to monitor and correct for changes that occur from differences between standards and samples. Since environmental samples often contain significant amounts of lithium, isotopically enriched 95% 6Li can be analyzed as an internal standard, avoiding the signal from the 7Li peak.