Claritas PPT® Grade ICP-MS Instrument Check Standard 3, SPEX CertiPrep
Catalog # SXCL-ICS-3
Supplier: SPEX CERTIPREP LLC
CAS Number:
Specifications
- Concentration:200 µg/ml
- Matrix:2% HNO₃
- Pack type:LDPE bottle
- Description:ICP-MS Instrument Check Standard 3
- Size:125 ml
- Cat. No.:SXCL-ICS-3
Specifications
About this item
Claritas PPT® multi-element standards for ICP-MS
SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.
The Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.
Certifications: SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.