ContactUsLinkComponent
Assurance® Grade Calibration Standards without Mercury for AA & ICP, SPEX CertiPrep
Assurance® Grade Calibration Standards without Mercury for AA & ICP, SPEX CertiPrep
Catalog # SXMXSTD-SETAN
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Assurance® Grade Calibration Standards without Mercury for AA & ICP, SPEX CertiPrep
Catalog # SXMXSTD-SETAN
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  MXSTD-SETAN
CAS Number:  
Specifications
  • Pack type:
    LDPE bottle
  • Description:
    Calibration Standards without Mercury
  • Size:
    5x 125 ml
  • Cat. No.:
    SXMXSTD-SETAN
Specifications
About this item
Assurance® mixed multi-element standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
AA & ICP Calibration Standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.
For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition).