Isolation Gowns, Medicom
76420-648
: Medicom
- Gender:Unisex
- Color:Yellow
- Description:X-Large
- Cat. No.:76420-648
- W×L:78×115 cm
Yellow isolation gowns are designed to protect against light fluid contact.
- Disposable design, single use only
- Two sizes available
- Latex-free
- Nonsterile
Gowns are constructed of a breathable fabric made from spunbond polypropylene. They feature sewn seams, elastic cuffs, adjustable neck ties and waist ties, with an opening in the back. The fluid repellent design provides added protection. Store in a cool, dry environment.
: Gowns are packaged 10 per bag, 5 bags per case.