Rhesus macaque Recombinant PLGF (from HEK293)
103738-728
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Size:50 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 100 mM Acetic Acid, pH2.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Rhesus macaque VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-R5257) at 5 μg/mL (100 μL/well) can bind Rhesus macaque PLGF, His Tag (Cat. No. PGF-R52H0) with a linear range of 2-40 ng/mL (QC tested).
- Gene ID:F7HB10-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:PGF|PLGF|PlGF2|PGFL|PlGF|SHGC-10760
- Protein/Peptide Name:PLGF
- Purity:>90%
- Molecular Weight:19.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 100 mM Acetic Acid, pH2.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103738-728