Rhesus macaque Recombinant OX40 Ligand (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Rhesus macaque OX40, His Tag (Cat. No. OX0-C5220) at 2 μg/mL (100 μL/well) can bind Rhesus macaque OX40 Ligand, Mouse IgG2a Fc Tag (Cat. No. OXL-R5259) with a linear range of 0.1-3 ng/mL (QC tested).
- Gene ID:F7FL80-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TNFSF4|CD134L|TXGP1|OX40L|Glycoprotein Gp34|CD134 ligand|CD252
- Protein/Peptide Name:OX40 Ligand
- Purity:>90%
- Molecular Weight:42.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together