Rhesus macaque Recombinant CD155 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Rhesus macaque CD155, His Tag (Cat. No. CD5-R52H5) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque TIGIT, Fc Tag (Cat. No. TIT-C5254) with a linear range of 0.02-0.313 μg/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Rhesus macaque CD155, His Tag (Cat. No. CD5-R52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human DNAM-1, Fc,Avitag (Cat. No. DN1-H82F9) with a linear range of 10-156 ng/mL (Routinely tested).; Loaded Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) on Protein A Biosensor, can bind Rhesus macaque CD155, His Tag (Cat. No. CD5-R52H5) with an affinity constant of 0.47 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q0MSE6-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
- Protein/Peptide Name:CD155
- Purity:>90%
- Molecular Weight:36.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together