Rat Recombinant CD47 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag (Cat. No. SIA-M5252) at 10 μg/mL (100 μL/well) can bind Rat CD47, Fc Tag (Cat. No. CD7-R5256) with a linear range of 2-20 ng/mL (QC tested).
- Gene ID:P97829
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IAP|MER6|OA3
- Protein/Peptide Name:CD47
- Purity:>95%
- Molecular Weight:40.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together