Rabbit Recombinant CTLA-4 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rabbit
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human B7-1, mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) at 5 μg/mL (100 μL/well)can bind Rabbit CTLA-4, Fc Tag (Cat. No. CT4-R5250) with a linear range of 0.1-0.8 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human B7-2, His Tag (Cat. No. CD6-H5223) at 5 μg/mL (100 μL/well)can bind Rabbit CTLA-4, Fc Tag (Cat. No. CT4-R5250) with a linear range of 5-78 ng/mL (Routinely tested).
- Gene ID:P42072
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CTLA4|CD152
- Protein/Peptide Name:CTLA-4
- Purity:>95%
- Molecular Weight:40.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together