Mouse Recombinant VEGF120 (from HEK293)
Catalog # 103737-762
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:25 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized VEGFR1/R2 at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His,Avitag (Cat. No. VE0-M82Q2) with a linear range of 0.1-2 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Avastin at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His,Avitag (Cat. No. VE0-M82Q2) with a linear range of 0.125-2 μg/mL (Routinely tested).
- Gene ID:Q00731-3
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:RP1-261G23.1|MVCD1|MGC70609|VEGFA|VPF
- Protein/Peptide Name:VEGF120
- Purity:>95%
- Molecular Weight:17.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103737-762