Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant VEGF R3 (from HEK293)
Mouse Recombinant VEGF R3 (from HEK293)
Catalog # 103738-144
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Mouse Recombinant VEGF R3 (from HEK293)
Catalog # 103738-144
Supplier:  ACROBIOSYSTEMS
Supplier Number:  FL4M5251100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 0.1 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human VEGF-C, His Tag (Cat. No. VEC-H4225) at 5 μg/mL (100 μL/well) can bind Mouse VEGF R3, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. FL4-M5251) with a linear range of 1-10 ng/mL (QC tested).
  • Gene ID:
    P35917-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Flt-4|VEGFR3|FLT4|LMPH1A|VEGFR-3|PCLFLT41|FLT-41|FLT41|FLT-4|PCL
  • Protein/Peptide Name:
    VEGF R3
  • Purity:
    >95%
  • Molecular Weight:
    111.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT
  • Cat. No.:
    103738-144

Specifications

About this item