Mouse Recombinant VEGF R2 (from HEK293)
Catalog # 103738-284
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human VEGF165, Tag Free (HPLC-verified) (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R2, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE2-M5258) with a linear range of 1-31 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Mouse VEGF120, Tag Free (Cat. No. VE0-M4211) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R2, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE2-M5258) with a linear range of 1-31 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Mouse VEGF164, Tag Free (HPLC-verified) (Cat. No. VE4-M4216) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R2, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE2-M5258) with a linear range of 2-31 ng/mL (Routinely tested).
- Gene ID:P35918-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:KDR|VEGFR|CD309|VEGFR2|FLK1
- Protein/Peptide Name:VEGF R2
- Purity:>95%
- Molecular Weight:110.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103738-284