Mouse Recombinant PD-L2 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human PD-1, Fc Tag (Cat. No. PD1-H82F1) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.2 μg/well) plate, can bind Mouse PD-L2, His Tag (Cat. No. PD2-M52E3) with a linear range of 20-156 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Mouse PD-L2, His Tag (Cat. No. PD2-M52E3) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.156-2.5 μg/mL (Routinely tested).; Loaded Mouse PD-1, Fc Tag (Cat. No. PD1-M5259) on Protein A Biosensor, can bind Mouse PD-L2, His Tag (Cat. No. PD2-M52E3) with an affinity constant of 0.13 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q9WUL5
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:PDCD1L2|CD273|PD-L2|Butyrophilin B7-DC|PDL2|PDCD1LG2|PDCD1 ligand 2
- Protein/Peptide Name:PD-L2
- Purity:>95%
- Molecular Weight:24.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together