Mouse Recombinant PD-L2 (from HEK293)
Marketsource
Supplier: ACRO Biosystems
About this item
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 103739-142
- 103739-144
- Protein/Peptide Name:
- PD-L2
- PD-L2
- Protein Synonyms:
- PDCD1L2|CD273|PD-L2|Butyrophilin B7-DC|PDL2|PDCD1LG2|PDCD1 ligand 2
- PDCD1L2|CD273|PD-L2|Butyrophilin B7-DC|PDL2|PDCD1LG2|PDCD1 ligand 2
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Source:
- HEK293 cells
- HEK293 cells
- Conjugation:
- Unconjugated
- Unconjugated
- Gene ID:
- Q9WUL5
- Q9WUL5
- Protease-free:
- Y
- Y
- Animal-Free:
- Y
- Y
- Nuclease-free:
- Y
- Y
- Carrier-Free:
- Y
- Y
- Endotoxin Level:
- Low
- Low
- Endotoxin Content:
- Less than 1.0 EU per μg by the LAL method.
- Less than 1.0 EU per μg by the LAL method.
- Biological Activity:
- Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse PD-L2, Fc Tag (Cat. No. PD2-M5254) at 5 μg/mL (100 μL/well) can bind Mouse PD-1, mouse IgG2a Fc tag (Cat. No. PD1-M52A4) with a linear range of 0.078-0.625 μg/mL (QC tested).
- Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse PD-L2, Fc Tag (Cat. No. PD2-M5254) at 5 μg/mL (100 μL/well) can bind Mouse PD-1, mouse IgG2a Fc tag (Cat. No. PD1-M52A4) with a linear range of 0.078-0.625 μg/mL (QC tested).
- Purity:
- >95%
- >95%
- Formulation:
- Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Molecular Weight:
- 49.2 kDa
- 49.2 kDa
- Storage Conditions:
- –20 °C
- –20 °C
- Shipping Temperature:
- Powder,RT
- Powder,RT
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: PD2M5254100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.103739-142Protein/Peptide NamePD-L2Protein SynonymsPDCD1L2|CD273|PD-L2|Butyrophilin B7-DC|PDL2|PDCD1LG2|PDCD1 ligand 2Protein/Peptide TypeRecombinantSpeciesMouseSourceHEK293 cellsConjugationUnconjugatedGene IDQ9WUL5Protease-freeYAnimal-FreeYNuclease-freeYCarrier-FreeYEndotoxin LevelLowEndotoxin ContentLess than 1.0 EU per μg by the LAL method.Biological ActivityLyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse PD-L2, Fc Tag (Cat. No. PD2-M5254) at 5 μg/mL (100 μL/well) can bind Mouse PD-1, mouse IgG2a Fc tag (Cat. No. PD1-M52A4) with a linear range of 0.078-0.625 μg/mL (QC tested).Purity>95%FormulationLyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.Molecular Weight49.2 kDaStorage Conditions–20 °CShipping TemperaturePowder,RTMarketsource - Supplier Number: PD2M52541MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.103739-144Protein/Peptide NamePD-L2Protein SynonymsPDCD1L2|CD273|PD-L2|Butyrophilin B7-DC|PDL2|PDCD1LG2|PDCD1 ligand 2Protein/Peptide TypeRecombinantSpeciesMouseSourceHEK293 cellsConjugationUnconjugatedGene IDQ9WUL5Protease-freeYAnimal-FreeYNuclease-freeYCarrier-FreeYEndotoxin LevelLowEndotoxin ContentLess than 1.0 EU per μg by the LAL method.Biological ActivityLyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse PD-L2, Fc Tag (Cat. No. PD2-M5254) at 5 μg/mL (100 μL/well) can bind Mouse PD-1, mouse IgG2a Fc tag (Cat. No. PD1-M52A4) with a linear range of 0.078-0.625 μg/mL (QC tested).Purity>95%FormulationLyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.Molecular Weight49.2 kDaStorage Conditions–20 °CShipping TemperaturePowder,RTMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



