Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant PD-L1 (from HEK293)
Mouse Recombinant PD-L1 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse Recombinant PD-L1 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 0.1 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse B7-1, Fc Tag (Cat. No. CD0-M5259) at 5 μg/mL (100 μL/well) can bind Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PD1-M52A2) with a linear range of 0.16-2.5 μg/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PD1-M52A2) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.313-5 μg/mL (Routinely tested).
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
  • Protein/Peptide Name:
    PD-L1
  • Purity:
    >95%
  • Molecular Weight:
    51.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together