Mouse Recombinant OX40 (from HEK293)
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse OX40 Ligand, His Tag (Cat. No. TN4-M5241) at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse OX40, His,Avitag (Cat. No. OX0-M82E5) with a linear range of 0.6-10 ng/mL (QC tested).
- Gene ID:P47741-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TXGP1L|ACT35|OX40|TNFRSF4|OX40L receptor|CD134
- Protein/Peptide Name:OX40
- Purity:>90%
- Molecular Weight:25.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together