Mouse Recombinant M-CSF R (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human M-CSF, His Tag (Cat. No. MCF-H5247) at 10 μg/mL (100 μL/well) can bind Biotinylated Mouse M-CSF R, Avitag,His Tag (Cat. No. CSR-M82E8) with a linear range of 0.625-10 μg/mL (QC tested).
- Gene ID:P09581-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD115|CSFR|M-CSFR|C-FMS|FMS|FIM2|CSF1R
- Protein/Peptide Name:M-CSF R
- Purity:>95%
- Molecular Weight:58.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together