Mouse Recombinant IL-17A (from HEK293)
Catalog # 103738-550
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) on Protein A Biosensor, can bind Mouse IL-17A, His Tag (Cat. No. CT8-M5240) with an affinity constant of 20.1 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Mouse IL-17A, His Tag (Cat. No. CT8-M5240) stimulates secretion of IL-6 by NIH-3T3 cells. The ED50 for this effect is 2.132-5.005 ng/mL.
- Gene ID:Q62386-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IL-17|IL-17A|CTLA-8|Interleukin-17A
- Protein/Peptide Name:IL-17A
- Purity:>90%
- Molecular Weight:15.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103738-550