Mouse Recombinant IgG3 Fc (from HEK293)
Catalog # 103739-026
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:200 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse IgG3 Fc, Tag Free (Cat. No. IG3-M5201) at 2 μg/mL (100 μL/well) can bind Human CD64, His Tag (Cat. No. FCA-H52H2) with a linear range of 2-20 ng/mL (QC tested).
- Gene ID:P03987-2
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IgG|IgG3|Ig gamma-3 chain C region
- Protein/Peptide Name:IgG3 Fc
- Purity:>95%
- Molecular Weight:26.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103739-026