Mouse Recombinant FGF R4 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse FGF R4, His Tag (Cat. No. FG4-M52Ha) at 2 μg/mL (100 μL/well) can bind Human FGF-9, Fc Tag (Cat. No. FG9-H4267) with a linear range of 0.156-5 μg/mL (QC tested).
- Gene ID:Q03142-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TKF|FGFR4|JTK2|CD334|MGC20292
- Protein/Peptide Name:FGF R4
- Purity:>90%
- Molecular Weight:40.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together