Order Entry
United States
ContactUsLinkComponent
 
Mouse Recombinant CD40 Ligand (from HEK293)
 
Mouse Recombinant CD40 Ligand (from HEK293)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse CD40, His Tag (Cat. No. TN5-M52H8) at 5 μg/mL (100 μL/well) can bind Mouse CD40 Ligand, Fc Tag (Cat. No. CDL-M526x) with a linear range of 1-16 ng/mL (QC tested).
  • Gene ID:
    P27548-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    TRAP|CD40L|IMD3|gp39|CD154|CD40LG|IGM|T-BAM|TNFSF5|HIGM1
  • Protein/Peptide Name:
    CD40 Ligand
  • Purity:
    >95%
  • Molecular Weight:
    42.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

 

Frequently Bought Together