Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant CD3 epsilon (from HEK293)
Mouse Recombinant CD3 epsilon (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse Recombinant CD3 epsilon (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P22646
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CD3E|TCRE|T3E|FLJ18683|CD3-epsilon
  • Protein/Peptide Name:
    CD3 epsilon
  • Purity:
    >95%
  • Molecular Weight:
    36 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together