Mouse Recombinant CD155 (from HEK293)
Catalog # 103739-316
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:50 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse TIGIT, His Tag (Cat. No. TIT-M52E6) at 10 μg/mL (100 μL/well) can bind Mouse CD155, Mouse IgG2a Fc Tag (Cat. No. CD5-M5254) with a linear range of 0.039-1.25 μg/mL (QC tested).
- Gene ID:Q8K094-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
- Protein/Peptide Name:CD155
- Purity:>95%
- Molecular Weight:61.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103739-316