Mouse Recombinant 4-1BB (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse 4-1BB, His Tag (Cat. No. 41B-M52H7) at 0.5 μg/mL (100 μL/well) can bind Mouse 4‑1BB Ligand, Fc Tag (Cat. No. 41L-M5257) with a linear range of 0.2-3 ng/mL (QC tested).; Loaded Mouse 4-1BB Ligand, Fc Tag (Cat. No. 41L-M5257) on Protein A Biosensor, can bind Mouse 4-1BB, His Tag (Cat. No. 41B-M52H7) with an affinity constant of 4.8 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:NP_001070976.1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD137|TNFRSF9|ILA|CDw137|4-1BB
- Protein/Peptide Name:4-1BB
- Purity:>95%
- Molecular Weight:21.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together