Order Entry
United States
ContactUsLinkComponent
Human Recombinant VEGF110 (from HEK293)
Human Recombinant VEGF110 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant VEGF110 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human VEGF110, Tag Free (HPLC-verified) (Cat. No. VE0-H5212) at 0.05 μg/mL (100 μL/well) can bind Human VEGF R1, His Tag (Cat. No. VE1-H5220) with a linear range of 4-63 ng/mL (QC tested).; ActiveMax® Human VEGF110, Tag Free (HPLC-verified) (Cat. No. VE0-H5212) stimulates proliferation of human umbilical vein endothelial cells (HUVEC). The EC50 for this effect is 5.06-11.59 ng/mL (Routinely tested).
  • Gene ID:
    P15692-4
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    RP1-261G23.1|MVCD1|MGC70609|VEGFA|VPF
  • Protein/Peptide Name:
    VEGF110
  • Purity:
    >95%
  • Molecular Weight:
    12.7 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together