Human Recombinant VEGF R3 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human VEGF-C, His Tag (Cat. No. VEC-H4225) at 5μg/mL (100 μL/well) can bind Human VEGF R3, Fc Tag (Cat. No. FL4-H5251) with a linear range of 0.6-10 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human VEGF R3, Fc Tag (Cat. No. FL4-H5251) at 5μg/mL (100 μL/well) can bind Human VEGF-D, His Tag (Cat. No. VED-H5228) with a linear range of 0.08-0.6 μg/mL (Routinely tested).
- Gene ID:P35916
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Flt-4|VEGFR3|FLT4|LMPH1A|VEGFR-3|PCLFLT41|FLT-41|FLT41|FLT-4|PCL
- Protein/Peptide Name:VEGF R3
- Purity:>95%
- Molecular Weight:111.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together