Human Recombinant TSLP R (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 μg/mL (100 μL/well) can bind Biotinylated Human TSLP, His,Avitag (Cat. No. TSP-H82Eb) with a linear range of 4-31 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Biotinylated Human TSLP (R127A, R130A), His,Avitag (Cat. No. TSP-H82E0) immobilized at 2 μg/mL (100 μL/well) via precoated 5 μg/mL (100 μL/well) of Human TSLP R, Fc Tag (Cat. No. TSR-H525a), can bind can bind Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. IL7-H5258) with a linear range of 1-20 ng/mL (Routinely tested).; Captured Human TSLP, His Tag (Cat. No. TSP-H52Hb) on CM5 Chip via anti-His antibody, can bind Human TSLP R, Fc Tag (Cat. No. TSR-H525a) with an affinity constant of 1.81 nM as determined in SPR assay (Biacore T200) (Routinely tested).; Captured Human TSLP (R127A, R130A), His Tag (Cat. No. TSP-H52Ha) on CM5 Chip via anti-His antibody, can bind Human TSLP R, Fc Tag (Cat. No. TSR-H525a) with an affinity constant of 4.22 nM as determined in SPR assay (Biacore T200) (Routinely tested).; Loaded Human TSLP R, Fc Tag (Cat. No. TSR-H525a) on Protein A Biosensor, can bind Human TSLP (R127A, R130A), His Tag (Cat. No. TSP-H52Ha) with an affinity constant of 16 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q9HC73-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TSLP receptor|CRL2|TSLPR|Cytokine receptor-like 2|ILXR|p2RY8/CRLF2 fusion|TSLP R|CRLF2|IL-XR|CRLF2Y
- Protein/Peptide Name:TSLP R
- Purity:>95%
- Molecular Weight:50.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together