Human Recombinant TSLP (from HEK293)
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 μg/mL (100 μL/well) can bind Human TSLP, His Tag (Cat. No. TSP-H52Hb) with a linear range of 1.6-13 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human TSLP, His Tag (Cat. No. TSP-H52Hb) at 1 μg/mL (100 μL/well) can bind Anti-TSLP MAb, Human IgG2 with a linear range of 0.5-8 ng/mL (Routinely tested).; Captured Human TSLP, His Tag (Cat. No. TSP-H52Hb) on CM5 Chip via anti-His antibody, can bind Human TSLP R, Fc Tag (Cat. No. TSR-H525a) with an affinity constant of 1.81 nM as determined in SPR assay (Biacore T200) (Routinely tested).
- Gene ID:Q969D9-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein/Peptide Name:TSLP
- Purity:>90%
- Molecular Weight:16.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together