Human Recombinant TNFSF11 (from HEK293)
Catalog # 103739-324
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Anti-TNFSF11 (human IgG1) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human TNFSF11, His Tag (active trimer) (Cat. No. RAL-H5240) with an affinity constant of 0.274 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Captured Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) on CM5 chip via Anti-Mouse antibodies surface can bind Human TNFSF11, His Tag (active trimer) (Cat. No. RAL-H5240) with an affinity constant of 0.309 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) on AMC Biosensor, can bind Human TNFSF11, His Tag (active trimer) (Cat. No. RAL-H5240) with an affinity constant of 0.409 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Anti-TNFSF11 MAb (human IgG1) on AHC Biosensor, can bind Human TNFSF11, His Tag (active trimer) (Cat. No. RAL-H5240) with an affinity constant of 0.239 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:AAC51762.1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:OPGL|ODF|TRANCE|CD254|RANKL
- Protein/Peptide Name:TNFSF11
- Purity:>90%
- Molecular Weight:22.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103739-324