Human Recombinant SLAMF7 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Humanized Anti-Human CS1 MAb at 1 μg/mL (100 μL/well) can bind Biotinylated Human SLAMF7, Avitag,His Tag (Cat. No. SL7-H82E0) with a linear range of 0.6-5 ng/mL (Routinely tested).; 2e5 of Anti-SLAMF7 CAR-293 cells were stained with 100 μL of 0.1 μg/mL of Biotinylated Human SLAMF7, Avitag, His Tag (Cat. No. SL7-H82E0) and negative control protein respectively, washed and then followed by PE-SA and analyzed with FACS (QC tested).
- Gene ID:Q9NQ25-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:FOAP-12|CRACC|19A|CS1|CD319|SLAMF7
- Protein/Peptide Name:SLAMF7
- Purity:>95%
- Molecular Weight:25.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together