Human Recombinant Siglec-2 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C, aviod light
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Monoclonal Anti-Human CD22 Antibody, Human IgG1 at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human Siglec-2, His Tag (Cat. No. SI2-HF2H6) with a linear range of 0.078-5 μg/mL (QC tested).; 293 cells were transfected with anti-CD22-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human Siglec-2, His Tag (Cat. No. SI2-HF2H6, 10 µg/mL) and C. FITC-labeled Protein Control. A. Non-transfected 293 cells and C. FITC-labeled Protein Control were used as negative control. RFP was used to evaluate CAR (anti-CD22-scFv) expression and FITC was used to evaluate the binding activity of FITC-Labeled Human Siglec-2, His Tag (Cat. No. SI2-HF2H6) (QC tested).
- Gene ID:P20273-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:BL-CAM|SIGLEC-2|CD22|Siglec2|SIGLEC2|SIGLEC2FLJ22814
- Protein/Peptide Name:Siglec-2
- Purity:>95%
- Molecular Weight:77.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT, Avoid light
Specifications
Frequently Bought Together