Human Recombinant PD-1 (from HEK293)
Catalog # 103738-006
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:1 mg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.01 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human PD-1, His Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H522a) at 2 μg/mL (100 μL/well) can bind Human PD-L2, Mouse IgG1 Fc Tag (Cat. No. PD2-H52A5) with a linear range of 10-156 ng/mL (Routinely tested).; Loaded Human PD-1, His Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H522a) on HIS1K Biosensor, can bind Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) with an affinity constant of 23.3 nM as determined in BLI assay (ForteBio Octet Red96e) (QC tested).; Loaded Human PD-1, His Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H522a) on HIS1K Biosensor, can bind Human PD-L2, Fc Tag (HPLC-verified) (Cat. No. PD2-H5251) with an affinity constant of 15.7 nM as determined in BLI assay (ForteBio Octet Red96e) (QC tested).; Loaded Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) on Protein A Biosensor, can bind Human Human PD-1, His Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H522a) with an affinity constant of 4.8 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human PD-L2, Fc Tag (HPLC verified) (Cat. No. PD2-H5251) on Protein A Biosensor, can bind Human Human PD-1, His Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H522a) with an affinity constant of 0.9 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q15116
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD279|PDCD1|PD1|SLEB2
- Protein/Peptide Name:PD-1
- Purity:>95%
- Molecular Weight:16.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103738-006