Human Recombinant Nectin-3 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Loaded Human Nectin-3, Fc Tag (Cat. No. PV3-H5255) on Protein A Biosensor, can bind Human TIGIT, His Tag (Cat. No. TIT-H52H3) with an affinity constant of 0.732 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:NP_056295.1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD113|Nectin-3|PVRL3
- Protein/Peptide Name:Nectin-3
- Purity:>95%
- Molecular Weight:64.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together