Human Recombinant Mesothelin (from HEK293), PE Labeled Protein
Catalog # 103737-662
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C, aviod light
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 10 mM Tris, 150 mM NaCl, 0.1% BSA, pH8.2. Normally trehalose is added as protectant before lyophilization.; 1e6 of the MSLN-CAR-293 cells were stained with 100 μl of 1:25 dilution (4 μL stock solution in 100 μl dilution buffer) of PE-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HP223). PE Streptavidin was used as negative control (QC tested).
- Gene ID:Q13421-3(R309P)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:MSLN,Mesothelin,MPF
- Protein/Peptide Name:Mesothelin
- Molecular Weight:33.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 10 mM Tris, 150 mM NaCl, 0.1% BSA, pH8.2. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT, Avoid light
- Cat. No.:103737-662