Human Recombinant Mesothelin (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C, aviod light
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human MSLN MAb at 10 μg/mL (100 μL/well) can bind GFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HG2P4) with a linear range of 0.039-0.625 μg/mL (QC tested).; Anti-human MSLN MAb (chimeric mouse-human IgG1) captured on CM5 chip via Anti-human IgG Fc antibodies surface, can bind GFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HG2P4) with an affinity constant of 0.542 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; 2e5 of the MSLN-CAR-293 cells were stained with 100 μL of 3 μg/mL of GFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HG2P4). GFP was used as negative control (Routinely tested).
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:MPF|MSLN|Mesothelin
- Protein/Peptide Name:Mesothelin
- Purity:>90%
- Molecular Weight:60.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT, Avoid light
Specifications
Frequently Bought Together