Order Entry
United States
ContactUsLinkComponent
Human Recombinant Mesothelin (from HEK293)
Human Recombinant Mesothelin (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant Mesothelin (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C, aviod light
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human MSLN MAb at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223) with a linear range of 0.078-0.313 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).; 293 cells were transfected with anti-MSLN-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223, 1 μg/mL) and C. FITC-labeled protein control. A. Non-transfected 293 cells and C. FITC-labeled protein control were used as negative control. RFP was used to evaluate CAR (anti-MSLN-scFv) expression and FITC was used to evaluate the binding activity of FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223).
  • Gene ID:
    Q13421-3(R309P)
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    MPF|MSLN|Mesothelin
  • Protein/Peptide Name:
    Mesothelin
  • Purity:
    >90%
  • Molecular Weight:
    33.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT, Avoid light

Specifications

Frequently Bought Together