Human Recombinant LILRA2 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRA2, His,Avitag (Cat. No. LI2-H82E9) with a linear range of 0.625-5 μg/mL (QC tested).
- Gene ID:Q8N149-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:ILT1CD85H|ILT-1|CD85h antigen|LIR-7|CD85H|CD85h|ILT1|LIR7|LILRA2|CD85 antigen-like family member H
- Protein/Peptide Name:LILRA2
- Purity:>90%
- Molecular Weight:50.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together