Human Recombinant Latent TGF-beta 1 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human ITGAV&ITGB6 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT6-H52E1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His,Avitag (Cat. No. TG1-H82Qb) with a linear range of 0.5-4 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His,Avitag (Cat. No. TG1-H82Qb) with a linear range of 0.2-4 ng/mL (Routinely tested).
- Gene ID:P01137-1 (C33S)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CED|TGF-beta-1|TGFB|DPD1|Latent TGF-beta 1|Latent TGFB1|LAP|TGFB1
- Protein/Peptide Name:Latent TGF-beta 1
- Purity:>90%
- Molecular Weight:45.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together