Human Recombinant Latent TGF-beta 1 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 1-20 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Human LRRC32, Fc Tag (Cat. No. LR2-H5256) with a linear range of 0.039-0.625 μg/mL (Routinely tested).; Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) inhibits the IL-4-dependent proliferation of TF-1 cells. The ED50 for this effect is 0.293-0.406 ng/mL (Routinely tested).
- Gene ID:P01137-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CED|TGF-beta-1|TGFB|DPD1|Latent TGF-beta 1|Latent TGFB1|LAP|TGFB1
- Protein/Peptide Name:Latent TGF-beta 1
- Purity:>95%
- Molecular Weight:42.6 kDa (monomer)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together